4-Bromo-o-cresol ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 4-Bromo-2-methylphenol, 1-Bromo-4-hydroxy-3-methylbenzene

001831-5G 001831-25G 001831-100G
101782-688EA 52.31 USD
101782-688 101782-690 101782-692
4-Bromo-o-cresol ≥98%
4-Bromo-o-cresol

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₇H₇BrO
MW: 187.04 g/mol
Melting Pt: 63.5 °C
MDL Number: MFCD00055435
CAS Number: 2362-12-1

Order Now

Specification Test Results

Melting point 63.5°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR