3-Bromo-o-toluic acid ≥98%

Supplier: Matrix Scientific

001820-25G 001820-100G
101782-674EA 58.46 USD
101782-674 101782-676
3-Bromo-o-toluic acid ≥98%
3-Bromo-o-toluic acid

Hazard code(s): Irritant

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Melting Pt: 154…156 °C
MDL Number: MFCD00270097
CAS Number: 76006-33-2

Order Now

Specification Test Results

Melting point 154-156°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR