2-Bromo-m-toluic acid ≥97%

Supplier: Matrix Scientific

001817-5G 001817-25G
101782-658EA 29.23 USD
101782-658 101782-660
2-Bromo-m-toluic acid ≥97%
2-Bromo-m-toluic acid

Hazard code(s): Irritant

Formula: C₈H₇BrO₂
Melting Pt: 134…136 °C
MDL Number: MFCD00079721
CAS Number: 53663-39-1

Order Now

Specification Test Results

Melting point 134-136°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR