2-Bromo-4-methylanisole ≥99%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
1-Bromo-2-methoxy-5-methylbenzene, 3-Bromo-4-methoxytoluene
001802-5G
001802-25G
101782-630EA
67.69
USD
101782-630
101782-632
2-Bromo-4-methylanisole ≥99%
2-Bromo-4-methylanisole
Hazard code(s): Irritant
Formula:
C₈H₉BrO MW: 201.06 g/mol Boiling Pt: 134 °C (25mm) Density: 1.41 g/cm³ |
MDL Number:
MFCD00017783 CAS Number: 22002-45-5 EINECS: 244-705-3 |
Specification Test Results
Boiling point | 134°C/25mm |
Density | 1.41 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...