(5-Bromo-2-methoxyphenyl)acetic acid ≥98%

Supplier: Matrix Scientific

001797-1G 001797-5G 001797-25G
101782-604EA 20 USD
101782-604 101782-606 101782-608
(5-Bromo-2-methoxyphenyl)acetic acid ≥98%
(5-Bromo-2-methoxyphenyl)acetic acid

Hazard code(s): Irritant

Formula: C₉H₉BrO₃
Melting Pt: 132…134 °C
MDL Number: MFCD00016825
CAS Number: 7017-48-3

Order Now

Specification Test Results

Melting point 132-134°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR