6-Bromo-m-anisaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-5-methoxybenzaldehyde

001791-5G 001791-25G 001791-100G
101782-582EA 29.23 USD
101782-582 101782-584 101782-586
6-Bromo-m-anisaldehyde ≥97%
6-Bromo-m-anisaldehyde

Hazard code(s): Irritant

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
MDL Number: MFCD00463817
CAS Number: 7507-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR