5-(Trifluoromethyl)-o-anisidine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methoxy-5-(trifluoromethyl)aniline, 5-(Trifluoromethyl)-2-anisidine, 5-(Trifluoromethyl)-2-methoxyaniline, 2-Amino-4-(trifluoromethyl)anisole, 3-Amino-4-methoxybenzotrifluoride

001790-5G 001790-25G
101782-578EA 23.08 USD
101782-578 101782-580
5-(Trifluoromethyl)-o-anisidine ≥97%
5-(Trifluoromethyl)-o-anisidine

Hazard code(s): Toxic

Formula: C₈H₈F₃NO
MW: 191.15 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 59 °C
MDL Number: MFCD00042486
CAS Number: 349-65-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 58-59°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR