2-Bromo-5-iodotoluene ≥97%

Supplier: Matrix Scientific
Warning

001784-5G 001784-25G
101782-570EA 30.77 USD
101782-570 101782-572
2-Bromo-5-iodotoluene ≥97%
2-Bromo-5-iodotoluene

Hazard code(s): Irritant, Light Sensitive

Formula: C₇H₆BrI
MW: 296.93 g/mol
Boiling Pt: 148 °C (20mm)
Density: 2.07 g/cm³
MDL Number: MFCD00070749
CAS Number: 202865-85-8

Order Now

Specification Test Results

Boiling point 148°C/20mm
Density 2.07

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR