3-Bromo-4-iodotoluene ≥99%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-1-iodo-4-methylbenzene, 1-Bromo-2-iodo-5-methylbenzene

001782-5G 001782-25G
101782-564EA 23.08 USD
101782-564 101782-566
3-Bromo-4-iodotoluene ≥99%
3-Bromo-4-iodotoluene

Hazard code(s): Irritant

Formula: C₇H₆BrI
MW: 296.93 g/mol
Boiling Pt: 133 °C (22mm)
Density: 2.08 g/cm³
MDL Number: MFCD00079718
CAS Number: 71838-16-9

Order Now

Specification Test Results

Boiling point 133°C/22mm
Density 2.08

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR