2-Bromo-5-iodobenzoic acid ≥99%

Supplier: Matrix Scientific
Warning

001777-1G 001777-5G
101782-552EA 15.38 USD
101782-552 101782-554
2-Bromo-5-iodobenzoic acid ≥99%
2-Bromo-5-iodobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄BrIO₂
MW: 326.92 g/mol
Melting Pt: 174…176 °C
MDL Number: MFCD00079716
CAS Number: 25252-00-0

Order Now

Specification Test Results

Melting point 174-176°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR