1-Bromo-3-iodobenzene ≥98%

Supplier: Matrix Scientific
Warning


Stabilization: Stabilised with copper

001776-25G 001776-100G
101782-548EA 29.23 USD
101782-548 101782-550
1-Bromo-3-iodobenzene ≥98%
1-Bromo-3-iodobenzene

(Stabilized with copper)


Hazard code(s): Irritant, Light Sensitive

Formula: C₆H₄BrI
Boiling Pt: 120 °C (18mm)
Melting Pt: –9 °C
Density: 2.21 g/cm³ (20 °C)
MDL Number: MFCD00001043
CAS Number: 591-18-4
EINECS: 209-703-9

Order Now

Specification Test Results

Boiling point 120°C/18mm
Melting point -9°C
Density 2.219
ND20 1.66


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR