5-Bromo-2-iodo-m-xylene ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-1,3-dimethyl-2-iodobenzene

001773-5G 001773-25G 001773-100G
101782-536EA 26.15 USD
101782-536 101782-538 101782-540
5-Bromo-2-iodo-m-xylene ≥97%
5-Bromo-2-iodo-m-xylene

Hazard code(s): Irritant

Formula: C₈H₈BrI
MW: 310.96 g/mol
Melting Pt: 42…44 °C
MDL Number: MFCD03094656
CAS Number: 206559-43-5

Order Now

Specification Test Results

Melting point 42-44°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR