5-Bromo-1-indanone ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2,3-dihydro-1H-inden-1-one

001772-25G 001772-100G
101782-532EA 66.15 USD
101782-532 101782-534
5-Bromo-1-indanone ≥98%
5-Bromo-1-indanone

Hazard code(s): Irritant

Formula: C₉H₇BrO
MW: 211.06 g/mol
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 125…127 °C
MDL Number: MFCD00082718
CAS Number: 34598-49-7

Order Now

Specification Test Results

Melting point 125-127°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR