4-Bromo-3-fluorotoluene ≥98%
Supplier: Matrix Scientific
|
Danger
|
001759-25G
001759-100G
101782-502EA
26.15
USD
101782-502
101782-504
4-Bromo-3-fluorotoluene ≥98%
4-Bromo-3-fluorotoluene
Hazard code(s): Irritant
Formula:
C₇H₆BrF Boiling Pt: 95 °C (50mm) Density: 1.494 g/cm³ |
MDL Number:
MFCD00040940 CAS Number: 452-74-4 UN: 1993 ADR: 3,III |
Specification Test Results
Boiling point | 95°C/50mm |
Density | 1.494 |
ND20 | 1.53 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...