4-Bromo-2-fluorotoluene ≥99%

Supplier: Matrix Scientific

001756-25G 001756-100G 001756-250G
101782-490EA 23.08 USD
101782-490 101782-492 101782-494
4-Bromo-2-fluorotoluene ≥99%
4-Bromo-2-fluorotoluene

Hazard code(s): Irritant

Formula: C₇H₆BrF
Boiling Pt: 68 °C (8mm)
Density: 1.5 g/cm³ (20 °C)
MDL Number: MFCD00013551
CAS Number: 51436-99-8
EINECS: 257-201-3

Order Now

Specification Test Results

Boiling point 68°C/8mm
Density 1.492
ND20 1.529


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR