2-Bromo-4-fluorotoluene ≥99%
Supplier: Matrix Scientific
|
Warning
|
001754-25G
001754-100G
101782-482EA
46.15
USD
101782-482
101782-484
2-Bromo-4-fluorotoluene ≥99%
2-Bromo-4-fluorotoluene
Hazard code(s): Irritant
Formula:
C₇H₆BrF Boiling Pt: 170…172 °C Density: 1.526 g/cm³ |
MDL Number:
MFCD00040828 CAS Number: 1422-53-3 EINECS: 215-830-0 |
Specification Test Results
Boiling point | 170-172°C |
Density | 1.526 |
ND20 | 1.528 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...