3-Bromo-4-fluorotoluene ≥99%

Supplier: Matrix Scientific
Warning

001753-25G 001753-100G
101782-478EA 21.54 USD
101782-478 101782-480
3-Bromo-4-fluorotoluene ≥99%
3-Bromo-4-fluorotoluene

Hazard code(s): Irritant

Formula: C₇H₆BrF
Boiling Pt: 169 °C
Density: 1.49 g/cm³
MDL Number: MFCD00040827
CAS Number: 452-62-0
EINECS: 207-201-4

Order Now

Specification Test Results

Boiling point 169°C
Density 1.49
ND20 1.531


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR