2-Amino-5-fluoropyridine ≥98%

Supplier: Matrix Scientific

001751-5G 001751-25G
101782-470EA 46.15 USD
101782-470 101782-472
2-Amino-5-fluoropyridine ≥98%
2-Amino-5-fluoropyridine

Hazard code(s): Irritant

Formula: C₅H₅FN₂
MW: 112.11 g/mol
Melting Pt: 94…96 °C
MDL Number: MFCD01861120
CAS Number: 21717-96-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 94-96°CC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR