4-Bromo-2-fluorophenol ≥98%

Supplier: Matrix Scientific
Warning

001747-25G 001747-100G
101782-466EA 27.69 USD
101782-466 101782-468
4-Bromo-2-fluorophenol ≥98%
4-Bromo-2-fluorophenol

Hazard code(s): Irritant

Formula: C₆H₄BrFO
Boiling Pt: 123…125 °C (50mm)
Density: 1.744 g/cm³
MDL Number: MFCD00011722
CAS Number: 2105-94-4

Order Now

Specification Test Results

Boiling point 123-125°C/5
Density 1.744
ND20 1.566


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR