4-Amino-3-fluorophenol ≥95%

Supplier: Matrix Scientific
Danger

001743-25G 001743-100G
101782-454EA 60 USD
101782-454 101782-456
4-Amino-3-fluorophenol ≥95%
4-Amino-3-fluorophenol

Hazard code(s): Irritant, Stored under Argon, Store in Freezer

Formula: C₆H₆FNO
MW: 127.12 g/mol
Melting Pt: 135…137 °C
MDL Number: MFCD00077452
CAS Number: 399-95-1
EINECS: 402-230-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Melting point 135-137°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR