1-Bromo-4-fluoronaphthalene ≥98%

Supplier: Matrix Scientific
Warning

001741-25G 001741-100G
101782-448EA 135.38 USD
101782-448 101782-450
1-Bromo-4-fluoronaphthalene ≥98%
1-Bromo-4-fluoronaphthalene

Hazard code(s): Irritant

Formula: C₁₀H₆BrF
MW: 225.06 g/mol
Melting Pt: 33…37 °C
MDL Number: MFCD00051473
CAS Number: 341-41-3
EINECS: 206-434-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR