2-Fluoro-5-(trifluoromethyl)aniline ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 3-Amino-alpha,alpha,alpha,4-tetrafluorotoluene, α,α,α,6-Tetrafluoro-m-toluidine, 3-Amino-α,α,α,4-tetrafluorotoluene, 3-Amino-4-fluorobenzotrifluoride

001734-25G 001734-100G
101782-438EA 26.15 USD
101782-438 101782-440
2-Fluoro-5-(trifluoromethyl)aniline ≥97%
2-Fluoro-5-(trifluoromethyl)aniline

Hazard code(s): Irritant

Formula: C₇H₅F₄N
MW: 179.12 g/mol
Boiling Pt: 81 °C (20mm)
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007653
CAS Number: 535-52-4
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 81°C/20mm
Density 1.378
ND20 1.461

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR