5-Bromo-α,α,α,2-tetrafluorotoluene ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-fluorobenzotrifluoride, 1-Bromo-4-fluoro-3-(trifluoromethyl)benzene

001733-25G 001733-100G
101782-434EA 33.85 USD
101782-434 101782-436
5-Bromo-α,α,α,2-tetrafluorotoluene ≥97%
5-Bromo-α,α,α,2-tetrafluorotoluene

Hazard code(s): Irritant

Formula: C₇H₃BrF₄
MW: 243 g/mol
Boiling Pt: 80 °C (50mm)
Density: 1.720 g/cm³
MDL Number: MFCD00042493
CAS Number: 393-37-3

Order Now

Specification Test Results

Boiling point 80°C/50mm
Density 1.72
ND20 1.46

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR