4-Fluoro-3-(trifluoromethyl)aniline ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 5-Amino-2-fluorobenzotrifluoride, 5-Amino-α,α,α,2-tetrafluorotoluene, α,α,α,4-Tetrafluoro-m-toluidine, 5-Amino-alpha,alpha,alpha,2-tetrafluorotoluene

001732-25G 001732-100G
101782-430EA 21.54 USD
101782-430 101782-432
4-Fluoro-3-(trifluoromethyl)aniline ≥98%
4-Fluoro-3-(trifluoromethyl)aniline

Hazard code(s): Toxic

Formula: C₇H₅F₄N
MW: 179.12 g/mol
Boiling Pt: 74…75 °C (10mm)
Density: 1.393 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00007834
CAS Number: 2357-47-3
EINECS: 219-095-7
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Boiling point 74-75°C/10m
Density 1.393


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR