3-Bromo-4-fluorobenzaldehyde ≥97%

Supplier: Matrix Scientific
Warning

001726-25G 001726-100G
101782-422EA 23.08 USD
101782-422 101782-424
3-Bromo-4-fluorobenzaldehyde ≥97%
3-Bromo-4-fluorobenzaldehyde

Hazard code(s): Irritant

Formula: C₇H₄BrFO
Boiling Pt: 138…139 °C (2.5mm)
Melting Pt: 29…32 °C
MDL Number: MFCD00042186
CAS Number: 77771-02-9
EINECS: 278-764-1

Order Now

Specification Test Results

Boiling point 138-139°C/2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR