4-Bromo-2-fluorobenzonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Brom-4-cyan-3-fluorbenzene

001724-25G 001724-100G
101782-414EA 69.23 USD
101782-414 101782-416
4-Bromo-2-fluorobenzonitrile ≥98%
4-Bromo-2-fluorobenzonitrile

Hazard code(s): Irritant

Formula: C₇H₃BrFN
MW: 200.01 g/mol
Boiling Pt: 238 - 239 °C (1013 hPa)
Melting Pt: 71 - 73 °C
MDL Number: MFCD00143264
CAS Number: 105942-08-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 68-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR