4-Bromo-3-fluorobenzoic acid ≥97%

Supplier: Matrix Scientific
Warning

001719-25G 001719-100G
101782-392EA 53.85 USD
101782-392 101782-394
4-Bromo-3-fluorobenzoic acid ≥97%
4-Bromo-3-fluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄BrFO₂
Melting Pt: 207…209 °C
MDL Number: MFCD00672932
CAS Number: 153556-42-4

Order Now

Specification Test Results

Melting point 207-209°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR