2-Bromo-6-fluorobenzoic acid ≥98% for synthesis
Supplier: Matrix Scientific
001715-5G
001715-25G
101782-376EA
35.38
USD
101782-376
101782-378
2-Bromo-6-fluorobenzoic acid ≥98% for synthesis
2-Bromo-6-fluorobenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₄BrFO₂ |
MDL Number:
MFCD01569539 CAS Number: 2252-37-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...