2-Bromo-5-fluorobenzoic acid ≥98%

Supplier: Matrix Scientific
Warning

001714-25G 001714-100G
101782-370EA 23.08 USD
101782-370 101782-372
2-Bromo-5-fluorobenzoic acid ≥98%
2-Bromo-5-fluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
Melting Pt: 151…153 °C
MDL Number: MFCD00142874
CAS Number: 394-28-5

Order Now

Specification Test Results

Melting point 151-153°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR