2-Bromo-3-fluorobenzoic acid ≥98%

Supplier: Matrix Scientific
Warning

001712-5G 001712-25G
101782-362EA 67.69 USD
101782-362 101782-364
2-Bromo-3-fluorobenzoic acid ≥98%
2-Bromo-3-fluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄BrFO₂
Melting Pt: 158…160 °C
MDL Number: MFCD01569398
CAS Number: 132715-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR