2-Amino-6-fluorobenzothiazole ≥99%

Supplier: Matrix Scientific

Synonyms: 6-Fluorobenzothiazol-2-ylamine, 6-Fluorobenzo[d]thiazol-2-amine, 6-Fluoro-1,3-benzothiazol-2-amine

001709-5G 001709-25G
101782-352EA 13.85 USD
101782-352 101782-354
2-Amino-6-fluorobenzothiazole ≥99%
2-Amino-6-fluorobenzothiazole

Hazard code(s): Irritant

Formula: C₇H₅FN₂S
MW: 168.19 g/mol
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 183…185 °C
Density: 1.59 g/cm³ (20 °C)
MDL Number: MFCD00013336
CAS Number: 348-40-3

Order Now

Specification Test Results

Melting point 183-185°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR