3-Bromo-α,α,α,2-tetrafluorotoluene ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-2-fluorobenzotrifluoride, 1-Bromo-2-fluoro-3-(trifluoromethyl)benzene

001703-1G 001703-5G
101782-336EA 23.08 USD
101782-336 101782-338
3-Bromo-α,α,α,2-tetrafluorotoluene ≥97%
3-Bromo-α,α,α,2-tetrafluorotoluene

Hazard code(s): Irritant

Formula: C₇H₃BrF₄
MW: 243 g/mol
MDL Number: MFCD00070812
CAS Number: 144584-67-8

Order Now

Specification Test Results

ND20 1.464

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR