5-Bromo-2-fluorobenzonitrile ≥97%

Supplier: Matrix Scientific

001700-5G 001700-25G 001700-100G
101782-322EA 23.08 USD
101782-322 101782-324 101782-326
5-Bromo-2-fluorobenzonitrile ≥97%
5-Bromo-2-fluorobenzonitrile

Hazard code(s): Irritant-Harmful

Formula: C₇H₃BrFN
MW: 200.01 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 75 - 80 °C
MDL Number: MFCD00143424
CAS Number: 179897-89-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 74-77°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR