3-Bromo-4-fluorobenzonitrile ≥98%

Supplier: Matrix Scientific

001699-25G 001699-100G
101782-318EA 52.31 USD
101782-318 101782-320
3-Bromo-4-fluorobenzonitrile ≥98%
3-Bromo-4-fluorobenzonitrile

Hazard code(s): Irritant-Harmful

Formula: C₇H₃BrFN
MW: 200.01 g/mol
Boiling Pt: 134 - 136 °C (33 -rr)
Melting Pt: 53 - 58 °C
MDL Number: MFCD00055432
CAS Number: 79630-23-2
EINECS: 279-200-7
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 53-55°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR