3-Fluoro-5-(trifluoromethyl)aniline ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 3-Amino-alpha,alpha,alpha,5-tetrafluorotoluene, 3-Amino-5-fluorobenzotrifluoride, 3-Amino-α,α,α,5-tetrafluorotoluene, α,α,α,5-Tetrafluoro-m-toluidine

001698-1G 001698-5G 001698-25G
101782-312EA 27.69 USD
101782-312 101782-314 101782-316
3-Fluoro-5-(trifluoromethyl)aniline ≥97% for synthesis
3-Fluoro-5-(trifluoromethyl)aniline

Hazard code(s): Irritant

Formula: C₇H₅F₄N
MW: 179.12
MDL Number: MFCD00061315
CAS Number: 454-67-1
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR