4-Fluoroanthranilic acid ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 2-Amino-4-fluorobenzoic acid

001695-25G 001695-100G
101782-302EA 41.54 USD
101782-302 101782-304
4-Fluoroanthranilic acid ≥98%
4-Fluoroanthranilic acid

Hazard code(s): Irritant

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 192…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00075553
CAS Number: 446-32-2
EINECS: 207-163-9

Order Now

Specification Test Results

Melting point 192-194°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR