4-Bromo-2-fluorobenzaldehyde ≥97%

Supplier: Matrix Scientific
Warning

001692-100G 001692-500G
101782-290EA 95.38 USD
101782-290 101782-292
4-Bromo-2-fluorobenzaldehyde ≥97%
4-Bromo-2-fluorobenzaldehyde

Hazard code(s): Irritant, Air Sensitive

Formula: C₇H₄BrFO
Boiling Pt: 42 °C (19mm)
Melting Pt: 60…62 °C
MDL Number: MFCD00143261
CAS Number: 57848-46-1

Order Now

Specification Test Results

Boiling point 42°C/19mm
Melting point 60-62°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR