4-Bromo-2-fluoroanisole ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 4-bromo-2-fluoro-1-methoxybenzene

001688-100G 001688-500G
101782-278EA 123.08 USD
101782-278 101782-280
4-Bromo-2-fluoroanisole ≥98%
4-Bromo-2-fluoroanisole

Hazard code(s): Irritant

Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: 16 °C
Density: 1.59 g/cm³ (20 °C)
MDL Number: MFCD00011710
CAS Number: 2357-52-0
EINECS: 219-096-2

Order Now

Specification Test Results

Boiling point 84°C/7mm
Density 1.545
ND20 1.59

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR