4-Bromo-2-fluoroaniline ≥98%

Supplier: Matrix Scientific
Warning

001685-25G 001685-100G
101782-274EA 23.08 USD
101782-274 101782-276
4-Bromo-2-fluoroaniline ≥98%
4-Bromo-2-fluoroaniline

Hazard code(s): Toxic, Refrigerate

Formula: C₆H₅BrFN
MW: 190.01 g/mol
Boiling Pt: 75 - 80 °C (0.3 -rr)
Melting Pt: 41 - 42 °C
MDL Number: MFCD00010221
CAS Number: 367-24-8
EINECS: 206-685-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 40-42°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR