5-Bromo-2-fluoro-m-xylene ≥99%

Supplier: Matrix Scientific
Warning

001683-1G 001683-5G
101782-270EA 36.92 USD
101782-270 101782-272
5-Bromo-2-fluoro-m-xylene ≥99%
5-Bromo-2-fluoro-m-xylene

Hazard code(s): Irritant

Formula: C₈H₈BrF
MW: 203.05 g/mol
Boiling Pt: 95 °C (22mm)
Density: 1.45 g/cm³
MDL Number: MFCD01320701
CAS Number: 99725-44-7

Order Now

Specification Test Results

Boiling point 95°C/22mm
Density 1.45

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR