1-Bromo-2-fluoro-4-iodobenzene ≥98%

Supplier: Matrix Scientific

001682-25G 001682-100G
101782-266EA 92.31 USD
101782-266 101782-268
1-Bromo-2-fluoro-4-iodobenzene ≥98%
1-Bromo-2-fluoro-4-iodobenzene

Hazard code(s): Irritant, Light Sensitive, Store in Freezer

Formula: C₆H₃BrFI
Boiling Pt: 80 °C (2.5mm)
Melting Pt: 37…38 °C
MDL Number: MFCD00042577
CAS Number: 136434-77-0

Order Now

Specification Test Results

Boiling point 80°C/2.5mm
Melting point 37-38°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR