1-Bromo-4-fluoro-3-nitrobenzene ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-1-fluoro-2-nitrobenzene, 5-Bromo-2-fluoronitrobenzene

001681-25G 001681-100G
101782-262EA 64.62 USD
101782-262 101782-264
1-Bromo-4-fluoro-3-nitrobenzene ≥97%
1-Bromo-4-fluoro-3-nitrobenzene

Hazard code(s): Irritant

Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Boiling Pt: 145 - 147 °C (35 -rr)
Melting Pt: 19 °C
Density: 1.786 g/cm³ (20 °C)
MDL Number: MFCD00129165
CAS Number: 364-73-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR