1-Bromo-4-fluoro-2-nitrobenzene ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Bromo-5-fluoronitrobenzene
001680-5G
001680-25G
101782-258EA
15.38
USD
101782-258
101782-260
1-Bromo-4-fluoro-2-nitrobenzene ≥97%
1-Bromo-4-fluoro-2-nitrobenzene
Hazard code(s): Irritant
Formula:
C₆H₃BrFNO₂ MW: 220 g/mol Melting Pt: 31 °C |
MDL Number:
MFCD00055530 CAS Number: 446-09-3 EINECS: 207-160-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Melting point | 31°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...