1-Bromo-3-fluoro-4-iodobenzene ≥95%

Supplier: Matrix Scientific
Warning

Synonyms: 3-Fluoro-4-iodobromobenzene, 4-Bromo-2-fluoro-1-iodobenzene

001677-25G 001677-100G
101782-252EA 38.46 USD
101782-252 101782-254
1-Bromo-3-fluoro-4-iodobenzene ≥95%
1-Bromo-3-fluoro-4-iodobenzene

Hazard code(s): Irritant, Refrigerate

Formula: C₆H₃BrFI
MW: 300.9 g/mol
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 48…51 °C
MDL Number: MFCD00010608
CAS Number: 105931-73-5

Order Now

Specification Test Results

Melting point 48-51°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR