1-Bromo-4-ethylbenzene ≥99%
Supplier: Matrix Scientific
Synonyms:
4-(Bromophenyl)ethane
001675-25G
001675-100G
101782-248EA
50.77
USD
101782-248
101782-250
1-Bromo-4-ethylbenzene ≥99%
1-Bromo-4-ethylbenzene
Hazard code(s): Irritant
Formula:
C₈H₉Br MW: 185.06 g/mol Boiling Pt: 204 °C Melting Pt: –43.5 °C Density: 1.34226 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000111 CAS Number: 1585-07-5 EINECS: 216-439-8 |
Specification Test Results
Boiling point | 204°C |
Density | 1.343 |
ND20 | 1.544 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...