1-Bromo-2-ethylbenzene ≥98%

Supplier: Matrix Scientific

001674-25G 001674-100G
101782-244EA 55.38 USD
101782-244 101782-246
1-Bromo-2-ethylbenzene ≥98%
1-Bromo-2-ethylbenzene

Hazard code(s): Irritant

Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 199 °C
Melting Pt: –67.3 °C
Density: 1.3589 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000077
CAS Number: 1973-22-4
EINECS: 217-823-8
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Boiling point 199°C
Density 1.35

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR