Ethyl-4,6-diphenylpyryliumtrifluoromethanesulfonate-2-carboxylate tech

Supplier: Matrix Scientific

001672-1G 001672-5G
101782-234EA 76.92 USD
101782-234 101782-236
Ethyl-4,6-diphenylpyryliumtrifluoromethanesulfonate-2-carboxylate tech
Ethyl 4,6-diphenylpyryliumtrifluoromethanesulphonate-2-carboxylate

Hazard code(s): Irritant

Formula: C₂₁H₁₇F₃O₆S
Melting Pt: 178…180 °C
MDL Number: MFCD00051895
CAS Number: 78904-85-5

Order Now

Specification Test Results

Melting point 178-180°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR