3-Bromo-4-chlorotoluene ≥99%

Supplier: Matrix Scientific

001656-5G 001656-25G
101782-208EA 15.38 USD
101782-208 101782-210
3-Bromo-4-chlorotoluene ≥99%
3-Bromo-4-chlorotoluene

Hazard code(s): Irritant, Light Sensitive

Formula: C₇H₆BrCl
Boiling Pt: 112 °C (22mm)
Density: 1.55 g/cm³
MDL Number: MFCD00079710
CAS Number: 57310-39-1

Order Now

Specification Test Results

Boiling point 112°C/22mm
Density 1.55


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR