4-Bromo-3-chlorotoluene ≥99%

Supplier: Matrix Scientific

001654-5G 001654-25G 001654-100G
101782-198EA 15.38 USD
101782-198 101782-200 101782-202
4-Bromo-3-chlorotoluene ≥99%
4-Bromo-3-chlorotoluene

Hazard code(s): Irritant

Formula: C₇H₆BrCl
Boiling Pt: 100 °C (22mm)
Density: 1.54 g/cm³
MDL Number: MFCD00045161
CAS Number: 6627-51-6

Order Now

Specification Test Results

Boiling point 100°C/22mm
Density 1.54


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR