2-Bromo-5-chlorothiophene ≥95%

Supplier: Matrix Scientific

001653-5G 001653-25G
101782-194EA 27.69 USD
101782-194 101782-196
2-Bromo-5-chlorothiophene ≥95%
2-Bromo-5-chlorothiophene

Hazard code(s): Irritant-Harmful

Formula: C₄H₂BrClS
MW: 197.48 g/mol
Boiling Pt: 69…70 °C (18mm)
Melting Pt: –2 °C
Density: 1.803 g/cm³ (25 °C)
MDL Number: MFCD00014523
CAS Number: 2873-18-9
EINECS: 220-708-5
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 69-70°C/18m
Density 1.803
ND20 1.596

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR